Learn More
Abnova™ Human RBL1 Partial ORF (AAH32247, 905 a.a. - 1014 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH32247 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5933 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124175
|
Abnova™
H00005933-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16114175
|
Abnova™
H00005933-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is similar in sequence and possibly function to the product of the retinoblastoma 1 (RB1) gene. The RB1 gene product is a tumor suppressor protein that appears to be involved in cell cycle regulation, as it is phosphorylated in the S to M phase transition and is dephosphorylated in the G1 phase of the cell cycle. Both the RB1 protein and the product of this gene can form a complex with adenovirus E1A protein and SV40 large T-antigen, with the SV40 large T-antigen binding only to the unphosphorylated form of each protein. In addition, both proteins can inhibit the transcription of cell cycle genes containing E2F binding sites in their promoters. Due to the sequence and biochemical similarities with the RB1 protein, it is thought that the protein encoded by this gene may also be a tumor suppressor. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: IDCDLEDATKTPDCSSGPVKEERGDLIKFYNTIYVGRVKSFALKYDLANQDHMMDAPPLSPFPHIKQQPGSPRRISQQHSIYISPHKNGSGLTPRSALLYKFNGSPSKVRSpecifications
AAH32247 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CP107/MGC40006/PRB1/p107 | |
RBL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5933 | |
RBL1 (Human) Recombinant Protein (Q01) | |
IDCDLEDATKTPDCSSGPVKEERGDLIKFYNTIYVGRVKSFALKYDLANQDHMMDAPPLSPFPHIKQQPGSPRRISQQHSIYISPHKNGSGLTPRSALLYKFNGSPSKVR | |
RUO | |
RBL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |