Learn More
Abnova™ Human Rax Partial Orf (Np_038463.2, 104 A.A.-206 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
Brand: Abnova™ H00030062-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: KEPWEARPSPGLPVGPATGEAKLSEEEQPKKKHRRNRTTFTTYQLHELERAFEKSHYPDVYSREELAGKVNLPEVRVQVWFQNRRAKWRRQEKLEVSSMKLQD
Specifications
NP_038463.2 | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
RAX (Human) Recombinant Protein (Q01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
KEPWEARPSPGLPVGPATGEAKLSEEEQPKKKHRRNRTTFTTYQLHELERAFEKSHYPDVYSREELAGKVNLPEVRVQVWFQNRRAKWRRQEKLEVSSMKLQD | |
RUO | |
RAX | |
GST |
Antibody Production, ELISA, Protein Array, Western Blot | |
30062 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MCOP3/RX | |
RAX | |
Liquid |
For Research Use Only