missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RAPGEFL1 Partial ORF (NP_057423, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051195-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106716

  • 349.00€ / 10µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

Sequence: MEGPEGLGRKQACLAMLLHFLDTYQGLLQEEEGAGHIIKDLYLLIMKDESLYQGLREDTLRLHQLVETVELKIPEENQPPSKQVKPLFRHFRRIDSCLQ
Specifications

Specifications

NP_057423
Liquid
51195
RAPGEFL1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Link-GEFII/MGC134798/MGC134799
RAPGEFL1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MEGPEGLGRKQACLAMLLHFLDTYQGLLQEEEGAGHIIKDLYLLIMKDESLYQGLREDTLRLHQLVETVELKIPEENQPPSKQVKPLFRHFRRIDSCLQ
RUO
RAPGEFL1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers