missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RAPGEF1 Partial ORF (AAH41710, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH41710 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2889 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115741
|
Abnova™
H00002889-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16125741
|
Abnova™
H00002889-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined. [provided by RefSeq]
Sequence: MGNAIEKQKPLKRSHLYPWKQDSQRSHLSSFTMKLMDKFHSPKIKRTPSKKGKPAEVSVKIPEKPVNKEATDRFLPEGYPLPLDLEQQAVEFMSTSAVASRSQRQKNLSWSpecifications
AAH41710 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C3G/DKFZp781P1719/GRF2 | |
RAPGEF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2889 | |
RAPGEF1 (Human) Recombinant Protein (Q01) | |
MGNAIEKQKPLKRSHLYPWKQDSQRSHLSSFTMKLMDKFHSPKIKRTPSKKGKPAEVSVKIPEKPVNKEATDRFLPEGYPLPLDLEQQAVEFMSTSAVASRSQRQKNLSW | |
RUO | |
RAPGEF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |