missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RAB19B Partial ORF (NP_001008749, 166 a.a. - 264 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00401409-Q01.25ug
This item is not returnable.
View return policy
Description
Sequence: GAANVVIMLIGNKCDLWEKRHVLFEDACTLAEKYGLLAVLETSAKESKNIEEVFVLMAKELIARNSLHLYGESALNGLPLDSSPVLMAQGPSEKTHCTCSpecifications
NP_001008749 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GAANVVIMLIGNKCDLWEKRHVLFEDACTLAEKYGLLAVLETSAKESKNIEEVFVLMAKELIARNSLHLYGESALNGLPLDSSPVLMAQGPSEKTHCTC | |
RUO | |
RAB19 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
401409 | |
RAB19B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RAB19B | |
RAB19 | |
Recombinant | |
wheat germ expression system |