missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human PTH1R / PTHR1 Protein, His Tag

Brand:  ACROBiosystems PTR-H5222-100UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15817567

  • 252.20€ / 100µg


Estimated Shipment: 20-10-2021
Add to basket

Description

Description

Parathyroid hormone/parathyroid hormone-related peptide receptor is also known as Parathyroid hormone 1 receptor (PTH1R), PTHR1, PTH/PTHrP type I receptor (PTH/PTHr receptor), which belongs to the G-protein coupled receptor 2 family. PTH1R / PTHR1 is a receptor for parathyroid hormone and for parathyroid hormone-related peptide. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.6 kDa. The protein migrates as 27-43 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Specifications

Specifications

NP_000307.1
HEK293
Human
19.6 kDa
100 µg
Asp 27 - Gly 188
PTH1R,PTHR,PTHR1
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>85% as determined by SDS-PAGE
Human PTH1R, His Tag (PTR-H5222) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Gly 188 (Accession # Q03431-1).
-20°C
PTH1R
His Tag
Videos
SDS
Documents
Certificates