missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTEN Partial ORF (NP_000305.1, 2 a.a. - 91 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005728-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16034615

  • 341.70€ / 10µg

Estimated Shipment: 24-01-2022
Add to basket

Description

Description

This gene was identified as a tumor suppressor that is mutated in a large number of cancers at high frequency. The protein encoded this gene is a phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase. It contains a tensin like domain as well as a catalytic domain similar to that of the dual specificity protein tyrosine phosphatases. Unlike most of the protein tyrosine phosphatases, this protein preferentially dephosphorylates phosphoinositide substrates. It negatively regulates intracellular levels of phosphatidylinositol-3,4,5-trisphosphate in cells and functions as a tumor suppressor by negatively regulating AKT/PKB signaling pathway. [provided by RefSeq]

Sequence: TAIIKEIVSRNKRRYQEDGFDLDLTYIYPNIIAMGFPAERLEGVYRNNIDDVVRFLDSKHKNHYKIYNLCAERHYDTAKFNCRVAQYPFE
Specifications

Specifications

NP_000305.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue
TAIIKEIVSRNKRRYQEDGFDLDLTYIYPNIIAMGFPAERLEGVYRNNIDDVVRFLDSKHKNHYKIYNLCAERHYDTAKFNCRVAQYPFE
RUO
PTEN
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
5728
PTEN (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
10q23del/BZS/MGC11227/MHAM/MMAC1/PTEN1/TEP1
PTEN
Yes
Videos
SDS
Documents
Certificates
Special Offers

Special Offers