missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PPP1R8 Partial ORF (NP_002704.1, 28 a.a. - 127 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_002704.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5511 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16103195
|
Abnova™
H00005511-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16193185
|
Abnova™
H00005511-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene, through alternative splicing, encodes three different isoforms. Two of the protein isoforms encoded by this gene are specific inhibitors of type 1 serine/threonine protein phosphatases and can bind but not cleave RNA. The third protein isoform lacks the phosphatase inhibitory function but is a single-strand endoribonuclease comparable to RNase E of E. coli. This isoform requires magnesium for its function and cleaves specific sites in A+U-rich regions of RNA. [provided by RefSeq]
Sequence: RRMQNFAFSGGLYGGLPPTHSEAGSQPHGIHGTALIGGLPMPYPNLAPDVDLTPVVPSAVNMNPAPNPAVYNPEAVNEPKKKKYAKEAWPGKKPTPSLLISpecifications
NP_002704.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARD-1/ARD1/NIPP-1/NIPP1/PRO2047 | |
PPP1R8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5511 | |
PPP1R8 (Human) Recombinant Protein (Q01) | |
RRMQNFAFSGGLYGGLPPTHSEAGSQPHGIHGTALIGGLPMPYPNLAPDVDLTPVVPSAVNMNPAPNPAVYNPEAVNEPKKKKYAKEAWPGKKPTPSLLI | |
RUO | |
PPP1R8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |