missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human POU2F3 Partial ORF (NP_055167, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00025833-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107802

  • 528.00€ / 25µg

Estimated Shipment: 12-07-2023
to see stock.
Add to basket

Description

Description

Sequence: MVNLESMHTDIKMSGDVADSTDARSTLSQVEPGNDRNGLDFNRQIKTEDLSDSLQQTLSHRPCHLSQGPAMMSGNQMSGLNASPCQDMASLHPLQQLVLVPGHLQSVSQ
Specifications

Specifications

NP_055167
Liquid
25833
POU2F3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Epoc-1/FLJ40063/MGC126698/OCT11/PLA-1/Skn-1a
POU2F3
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MVNLESMHTDIKMSGDVADSTDARSTLSQVEPGNDRNGLDFNRQIKTEDLSDSLQQTLSHRPCHLSQGPAMMSGNQMSGLNASPCQDMASLHPLQQLVLVPGHLQSVSQ
RUO
POU2F3
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers