Learn More
Abnova™ Human POMT2 Partial ORF (NP_037514, 483 a.a. - 580 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029954-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT1 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS)
Sequence: CVLGSSGKVLPKWGWEQLEVTCTPYLKETLNSIWNVEDHINPKLPNISLDVLQPSFPEILLESHMVMIRGNSGLKPKDNEFTSKPWHWPINYQGLRFSSpecifications
NP_037514 | |
Liquid | |
29954 | |
POMT2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686G10254/FLJ22309 | |
POMT2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CVLGSSGKVLPKWGWEQLEVTCTPYLKETLNSIWNVEDHINPKLPNISLDVLQPSFPEILLESHMVMIRGNSGLKPKDNEFTSKPWHWPINYQGLRFS | |
RUO | |
POMT2 | |
Wheat Germ (in vitro) | |
GST |