missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PLCD4 Full-length ORF (AAH06355, 1 a.a. - 762 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH06355 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84812 |
Molecular Weight (g/mol) | 109.34kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16156083
|
Abnova™
H00084812-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16146083
|
Abnova™
H00084812-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Phosphatidylinositol-specific phospholipase C (PLC) plays an important role in receptor-mediated signal transduction by generating 2 second messenger molecules, inositol 1,4,5-triphosphate (IP3) and diacylglycerol, from phosphatidylinositol 4,5-bisphosphate (PIP2). PLC comprises a diverse family of enzymes that differ in structure and tissue distribution (Berridge, 1993 [PubMed 8381210]).[supplied by OMIM]
Sequence: MASLLQDQLTTDQDLLLMQEGMPMRKVRSKSWKKLRYFRLQNDGMTVWHARQARGSAKPSFSISDVETIRNGHDSELLRSLAEELPLEQGFTIVFHGRRSNLDLMANSVGEAQIWMRGLQLLVDLVTSMDHQERLDQWLSDWFQRGDKNQDGKMSFQEVQRLLHLMNVEMDQEYAFSLFQAADTSQSGTLEGEEFVQFYKALTKRAEVQELFESFSADGQKLTLLEFLDFLQEEQKERDCTSELALELIDRYEPSDSGKLRHVLSMDGFLSYLCSKDGDIFNPACLPIYQDMTQPLNHYFICSSHNTYLVGDQLCGQSSVEGYIRALKRGCRCVEVDVWDGPSGEPVVYHGHTLTSRILFKDVVATVAQYAFQTSDYPVILSLETHCSWEQQQTMARHLTEILGEQLLSTTLDGVLPTQLPSPEELRRKILVKGKKLTLEEDLEYEEEEAEPELEESELALESQFETEPEPQEQNLQNKDKKKKSKPILCPALSSLVIYLKSVSFRSFTHSKEHYHFYEISSFSETKAKRLIKEAGNEFVQHNTWQLSRVYPSGLRTDSSNYNPQELWNAGCQMVAMNMQTAGLEMDICDGHFRQNGGCGYVLKPDFLRDIQSSFHPEKPISPFKAQTLLIQVISGQQLPKVDKTKEGSIVDPLVKVQIFGVRLDTARQETNYVENNGFNPYWGQTLCFRVLVPELAMLRFVVMDYDWKSRNDFIGQYTLPWTCMQQGYRHIHLLSKDGISLRPASIFVYICIQEGLEGDESSpecifications
AAH06355 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109.34kDa | |
Glutathione Sepharose 4 Fast Flow | |
MASLLQDQLTTDQDLLLMQEGMPMRKVRSKSWKKLRYFRLQNDGMTVWHARQARGSAKPSFSISDVETIRNGHDSELLRSLAEELPLEQGFTIVFHGRRSNLDLMANSVGEAQIWMRGLQLLVDLVTSMDHQERLDQWLSDWFQRGDKNQDGKMSFQEVQRLLHLMNVEMDQEYAFSLFQAADTSQSGTLEGEEFVQFYKALTKRAEVQELFESFSADGQKLTLLEFLDFLQEEQKERDCTSELALELIDRYEPSDSGKLRHVLSMDGFLSYLCSKDGDIFNPACLPIYQDMTQPLNHYFICSSHNTYLVGDQLCGQSSVEGYIRALKRGCRCVEVDVWDGPSGEPVVYHGHTLTSRILFKDVVATVAQYAFQTSDYPVILSLETHCSWEQQQTMARHLTEILGEQLLSTTLDGVLPTQLPSPEELRRKILVKGKKLTLEEDLEYEEEEAEPELEESELALESQFETEPEPQEQNLQNKDKKKKSKPILCPALSSLVIYLKSVSFRSFTHSKEHYHFYEISSFSETKAKRLIKEAGNEFVQHNTWQLSRVYPSGLRTDSSNYNPQELWNAGCQMVAMNMQTAGLEMDICDGHFRQNGGCGYVLKPDFLRDIQSSFHPEKPISPFKAQTLLIQVISGQQLPKVDKTKEGSIVDPLVKVQIFGVRLDTARQETNYVENNGFNPYWGQTLCFRVLVPELAMLRFVVMDYDWKSRNDFIGQYTLPWTCMQQGYRHIHLLSKDGISLRPASIFVYICIQEGLEGDES | |
RUO | |
PLCD4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84812 | |
PLCD4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC12837 | |
PLCD4 | |
Recombinant | |
wheat germ expression system |