missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLA2G10 Full-length ORF (NP_003552.1, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008399-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192412

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MGPLPVCLPIMLLLLLPSLLLLLLLPGPGSGEASRILRVHRRGILELAGTVGCVGPRTPIAYMKYGCFCGLGGHGQPRDAIDWCCHGHDCCYTRAEEAGCSPKTERYSWQCVNQSVLCGPAENKCQELLCKCDQEIANCLAQTEYNLKYLFYPQFLCEPDSPKCD
Specifications

Specifications

NP_003552.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GXPLA2/GXSPLA2/MGC119918/MGC119919/MGC133367/SPLA2
PLA2G10
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8399
PLA2G10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGPLPVCLPIMLLLLLPSLLLLLLLPGPGSGEASRILRVHRRGILELAGTVGCVGPRTPIAYMKYGCFCGLGGHGQPRDAIDWCCHGHDCCYTRAEEAGCSPKTERYSWQCVNQSVLCGPAENKCQELLCKCDQEIANCLAQTEYNLKYLFYPQFLCEPDSPKCD
RUO
PLA2G10
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers