missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PITPNC1 Full-length ORF (AAH07905, 1 a.a. - 268 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026207-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16128172

  • 508.00€ / 25µg
Estimated Shipment: 07-05-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the phosphatidylinositol transfer protein family. The encoded cytoplasmic protein transfers phosphatidylinositol from one membrane compartment to another. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MLLKEYRICMPLTVDEYKIGQLYMISKHSHEQSDRGEGVEVVQNEPFEDPHHGNGQFTEKRVYLNSKLPSWARAVVPKIFYVTEKAWNYYPYTITEYTCSFLPRFSIHIETKYEDNKGSNDTIFDNEAKDVEREVCFIDIACDEIPERYYKESEDPKHFKSEKTGRGQLREGWRDSHQPIMCSYKLVTVKFEVWGLQTRVEQFVHKVVRDILLIGHRQAFAWVDEWYDMTMDDVREYEKNMHEQTNIKVCNQHSSPVDDIESHAQTST
Specifications

Specifications

AAH07905
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.22kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RDGB-BETA/RDGBB/RDGBB1
PITPNC1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
26207
PITPNC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLLKEYRICMPLTVDEYKIGQLYMISKHSHEQSDRGEGVEVVQNEPFEDPHHGNGQFTEKRVYLNSKLPSWARAVVPKIFYVTEKAWNYYPYTITEYTCSFLPRFSIHIETKYEDNKGSNDTIFDNEAKDVEREVCFIDIACDEIPERYYKESEDPKHFKSEKTGRGQLREGWRDSHQPIMCSYKLVTVKFEVWGLQTRVEQFVHKVVRDILLIGHRQAFAWVDEWYDMTMDDVREYEKNMHEQTNIKVCNQHSSPVDDIESHAQTST
RUO
PITPNC1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers