Learn More
Abnova™ Human PIGW Partial ORF (NP_848612.2, 399 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00284098-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Glycosylphosphatidylinositol (GPI) is a complex glycolipid that anchors many proteins to the cell surface. PIGW acts in the third step of GPI biosynthesis and acylates the inositol ring of phosphatidylinositol (Murakami et al., 2003 [PubMed 14517336]).[supplied by OMIM]
Sequence: KFLIKGALVPCSWKLIQSPVTNKKHSESLVPEAERMEPSLCLITALNRKSpecifications
NP_848612.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.13kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KFLIKGALVPCSWKLIQSPVTNKKHSESLVPEAERMEPSLCLITALNRK | |
RUO | |
PIGW | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
284098 | |
PIGW (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ37433/Gwt1 | |
PIGW | |
Recombinant | |
wheat germ expression system |