Learn More
Abnova™ Human PGBD1 Full-length ORF (NP_115896.1, 1 a.a. - 809 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_115896.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84547 |
Molecular Weight (g/mol) | 118.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115953
|
Abnova™
H00084547-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16105953
|
Abnova™
H00084547-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene product is specifically expressed in the brain, however, its exact function is not known. [provided by RefSeq]
Sequence: MYEALPGPAPENEDGLVKVKEEDPTWEQVCNSQEGSSHTQEICRLRFRHFCYQEAHGPQEALAQLRELCHQWLRPEMHTKEQIMELLVLEQFLTILPKELQPCVKTYPLESGEEAVTVLENLETGSGDTGQQASVYIQGQDMHPMVAEYQGVSLECQSLQLLPGITTLKCEPPQRPQGNPQEVSGPVPHGSAHLQEKNPRDKAVVPVFNPVRSQTLVKTEEETAQAVAAEKWSHLSLTRRNLCGNSAQETVMSLSPMTEEIVTKDRLFKAKQETSEEMEQSGEASGKPNRECAPQIPCSTPIATERTVAHLNTLKDRHPGDLWARMHISSLEYAAGDITRKGRKKDKARVSELLQGLSFSGDSDVEKDNEPEIQPAQKKLKVSCFPEKSWTKRDIKPNFPSWSALDSGLLNLKSEKLNPVELFELFFDDETFNLIVNETNNYASQKNVSLEVTVQEMRCVFGVLLLSGFMRHPRREMYWEVSDTDQNLVRDAIRRDRFELIFSNLHFADNGHLDQKDKFTKLRPLIKQMNKNFLLYAPLEEYYCFDKSMCECFDSDQFLNGKPIRIGYKIWCGTTTQGYLVWFEPYQEESTMKVDEDPDLGLGGNLVMNFADVLLERGQYPYHLCFDSFFTSVKLLSALKKKGVRATGTIRENRTEKCPLMNVEHMKKMKRGYFDFRIEENNEIILCRWYGDGIISLCSNAVGIEPVNEVSCCDADNEEIPQISQPSIVKVYDECKEGVAKMDQIISKYRVRIRSKKWYSILVSYMIDVAMNNAWQLHRACNPGASLDPLDFRRFVAHFYLEHNAHLSDSpecifications
NP_115896.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MYEALPGPAPENEDGLVKVKEEDPTWEQVCNSQEGSSHTQEICRLRFRHFCYQEAHGPQEALAQLRELCHQWLRPEMHTKEQIMELLVLEQFLTILPKELQPCVKTYPLESGEEAVTVLENLETGSGDTGQQASVYIQGQDMHPMVAEYQGVSLECQSLQLLPGITTLKCEPPQRPQGNPQEVSGPVPHGSAHLQEKNPRDKAVVPVFNPVRSQTLVKTEEETAQAVAAEKWSHLSLTRRNLCGNSAQETVMSLSPMTEEIVTKDRLFKAKQETSEEMEQSGEASGKPNRECAPQIPCSTPIATERTVAHLNTLKDRHPGDLWARMHISSLEYAAGDITRKGRKKDKARVSELLQGLSFSGDSDVEKDNEPEIQPAQKKLKVSCFPEKSWTKRDIKPNFPSWSALDSGLLNLKSEKLNPVELFELFFDDETFNLIVNETNNYASQKNVSLEVTVQEMRCVFGVLLLSGFMRHPRREMYWEVSDTDQNLVRDAIRRDRFELIFSNLHFADNGHLDQKDKFTKLRPLIKQMNKNFLLYAPLEEYYCFDKSMCECFDSDQFLNGKPIRIGYKIWCGTTTQGYLVWFEPYQEESTMKVDEDPDLGLGGNLVMNFADVLLERGQYPYHLCFDSFFTSVKLLSALKKKGVRATGTIRENRTEKCPLMNVEHMKKMKRGYFDFRIEENNEIILCRWYGDGIISLCSNAVGIEPVNEVSCCDADNEEIPQISQPSIVKVYDECKEGVAKMDQIISKYRVRIRSKKWYSILVSYMIDVAMNNAWQLHRACNPGASLDPLDFRRFVAHFYLEHNAHLSD | |
RUO | |
PGBD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84547 | |
PGBD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUCEP-4/SCAND4/dJ874C20.4 | |
PGBD1 | |
Recombinant | |
wheat germ expression system |