Learn More
ACROBiosystems Human PD-1 / PDCD1 Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
457.25€ - 3357.52€
Specifications
Accession Number | NP 005009.2 |
---|---|
Molecular Weight (g/mol) | 43.8 kDa |
Source | Human PD-1, Llama IgG2b Fc Tag, low endotoxin (PD1-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1). |
Immunogen | Leu 25 - Gln 167 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15817467
|
ACROBiosystems
PD1-H5259-100UG |
100 μg |
457.25€
100µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
15827467
|
ACROBiosystems
PD1-H5259-1MG |
1 mg |
3357.52€
1mg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.
This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 43.8 kDa. The protein migrates as 55-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP 005009.2 | |
Human PD-1, Llama IgG2b Fc Tag, low endotoxin (PD1-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1). | |
-20°C | |
PD-1 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |
43.8 kDa | |
Leu 25 - Gln 167 | |
PDCD1,PD1,CD279,SLEB2 | |
Unconjugated | |
Llama IgG Fc Tag | |
Less than 0.01 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |