missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PCYT1B Partial ORF (NP_004836, 236 a.a. - 314 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004836 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9468 |
Molecular Weight (g/mol) | 34.43kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100746
|
Abnova™
H00009468-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16190736
|
Abnova™
H00009468-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene belongs to the cytidylyltransferase family. It is involved in the regulation of phosphatidylcholine biosynthesis. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: NEKRYRFQNQVDKMKEKVKNVEERSKEFVNRVEEKSHDLIQKWEEKSREFIGNFLELFGPDGAWKQMFQERSSRMLQALSpecifications
NP_004836 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.43kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCT-beta/CTB | |
PCYT1B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9468 | |
PCYT1B (Human) Recombinant Protein (Q01) | |
NEKRYRFQNQVDKMKEKVKNVEERSKEFVNRVEEKSHDLIQKWEEKSREFIGNFLELFGPDGAWKQMFQERSSRMLQAL | |
RUO | |
PCYT1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |