Learn More
Abnova™ Human PCGF6 Partial ORF (NP_001011663, 225 a.a. - 324 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001011663 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84108 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16120637
|
Abnova™
H00084108-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16110637
|
Abnova™
H00084108-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene contains a RING finger motif, which is most closely related to those of polycomb group (PcG) proteins RNF110/MEL-18 and BMI1. PcG proteins are known to form protein complexes and function as transcription repressors. This protein has been shown to interact with some PcG proteins and act as a transcription repressor. The activity of this protein is found to be regulated by cell cycle dependent phosphorylation. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: AVPQPVPSSKGRSKKVLESVFRIPPELDMSLLLEFIGANEGTGHFKPLEKKFVRVSGEATIGHVEKFLRRKMGLDPACQVDIICGDHLLEQYQTLREIRRSpecifications
NP_001011663 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MBLR/MGC15678/MGC17541/RNF134 | |
PCGF6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
84108 | |
PCGF6 (Human) Recombinant Protein (Q01) | |
AVPQPVPSSKGRSKKVLESVFRIPPELDMSLLLEFIGANEGTGHFKPLEKKFVRVSGEATIGHVEKFLRRKMGLDPACQVDIICGDHLLEQYQTLREIRR | |
RUO | |
PCGF6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |