missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PCGF3 Partial ORF (NP_006306, 133 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_006306 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10336 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16174912
|
Abnova™
H00010336-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16164912
|
Abnova™
H00010336-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene contains a C3HC4 type RING finger, which is a motif known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined. [provided by RefSeq]
Sequence: SNKEAAEEKPEEDNDYHRSDEQVSICLECNSSKLRGLKRKWIRCSAQATVLHLKKFIAKKLNLSSFNELDILCNEEILGKDHTLKFVVVTRWRFKKAPLLLHYRPKMDLLSpecifications
NP_006306 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686D20235/DONG1/FLJ36550/FLJ43813/MGC129615/MGC40413/RNF3/RNF3A | |
PCGF3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10336 | |
PCGF3 (Human) Recombinant Protein (Q01) | |
SNKEAAEEKPEEDNDYHRSDEQVSICLECNSSKLRGLKRKWIRCSAQATVLHLKKFIAKKLNLSSFNELDILCNEEILGKDHTLKFVVVTRWRFKKAPLLLHYRPKMDLL | |
RUO | |
PCGF3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |