missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PAX3 Partial ORF (NP_000429, 84 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005077-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122385

  • 508.00€ / 25µg
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

This gene is a member of the paired box (PAX) family of transcription factors. Members of the PAX family typically contain a paired box domain and a paired-type homeodomain. These genes play critical roles during fetal development. Mutations in paired box gene 3 are associated with Waardenburg syndrome, craniofacial-deafness-hand syndrome, and alveolar rhabdomyosarcoma. The translocation t(2;13)(q35;q14), which represents a fusion between PAX3 and the forkhead gene, is a frequent finding in alveolar rhabdomyosarcoma. Alternative splicing results in transcripts encoding isoforms with different C-termini. [provided by RefSeq]

Sequence: SKILCRYQETGSIRPGAIGGSKPKQVTTPDVEKKIEEYKRENPGMFSWEIRDKLLKDAVCDRNTVPSVSSISRILRSKFGKG
Specifications

Specifications

NP_000429
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.76kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SKILCRYQETGSIRPGAIGGSKPKQVTTPDVEKKIEEYKRENPGMFSWEIRDKLLKDAVCDRNTVPSVSSISRILRSKFGKG
RUO
PAX3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5077
PAX3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDHS/HUP2/MGC120381/MGC120382/MGC120383/MGC120384/MGC134778/WS1
PAX3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers