missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OXCT1 Partial ORF (NP_000427, 75 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005019-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192265

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a member of the 3-oxoacid CoA-transferase gene family. The encoded protein is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate. Mutations in this gene are associated with succinyl CoA:3-oxoacid CoA transferase deficiency. [provided by RefSeq]

Sequence: DALLKTGVKGLTAVSNNAGVDNFGLGLLLRSKQIKRMVSSYVGENAEFERQYLS
Specifications

Specifications

NP_000427
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.68kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DALLKTGVKGLTAVSNNAGVDNFGLGLLLRSKQIKRMVSSYVGENAEFERQYLS
RUO
OXCT1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5019
OXCT1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OXCT/SCOT
OXCT1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers