missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR2A2 Partial ORF (NP_001005480.1, 261 a.a. - 318 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00442361-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192997

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: PDSNQREEQEKMLSLFHSVLNPMLNPLIYSLRNAQLKGALHRALQRKRSMRTVYGLCL
Specifications

Specifications

NP_001005480.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.12kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PDSNQREEQEKMLSLFHSVLNPMLNPLIYSLRNAQLKGALHRALQRKRSMRTVYGLCL
RUO
OR2A2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
442361
OR2A2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OR2A17P/OR2A2P/OR7-11/OST008
OR2A2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers