Learn More
Abnova™ Human NPHP4 Partial ORF (NP_055917.1, 1317 a.a. - 1426 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00261734-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which contains a proline-rich region. The encoded protein may function in renal tubular development and function. This protein interacts with nephrocystin. Mutations in this gene are associated with nephronophthisis type 4. Multiple alternative transcript variants have been described but their full-length nature has not been determined. [provided by RefSeq]
Sequence: ASWLVCLCCRQPLISKAFEIMLAAGEGKGVNKRITYTNPYPSRRTFHLHSDHPELLRFREDSFQVGGGETYTIGLQFAPSQRVGEEEILIYINDHEDKNEEAFCVKVIYQSpecifications
NP_055917.1 | |
Liquid | |
261734 | |
NPHP4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0673/SLSN4 | |
NPHP4 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ASWLVCLCCRQPLISKAFEIMLAAGEGKGVNKRITYTNPYPSRRTFHLHSDHPELLRFREDSFQVGGGETYTIGLQFAPSQRVGEEEILIYINDHEDKNEEAFCVKVIYQ | |
RUO | |
NPHP4 | |
Wheat Germ (in vitro) | |
GST |