missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NOXO1 Partial ORF (AAH15917, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH15917 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 124056 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101677
|
Abnova™
H00124056-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16191667
|
Abnova™
H00124056-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
NADPH oxidases (NOXs) catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species (ROS). NOX organizers, such as NOXO1, target NOX activators (see NOXA1; MIM 611255) to NOX and also target NOX to different subcellular compartments (Opitz et al., 2007 [PubMed 17189823]).[supplied by OMIM]
Sequence: MAGPRYPVSVQGAALVQIKRLQTFAFSVRWSDGSDTFVRRSWDEFRQLKTLKETFPVEAGLLRRSDRVLPKLLDAPLLGRVGRTSRGLARLQLLETYSRRSpecifications
AAH15917 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC20258/P41NOX/P41NOXA/P41NOXB/P41NOXC/SH3PXD5/SNX28 | |
NOXO1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
124056 | |
NOXO1 (Human) Recombinant Protein (Q01) | |
MAGPRYPVSVQGAALVQIKRLQTFAFSVRWSDGSDTFVRRSWDEFRQLKTLKETFPVEAGLLRRSDRVLPKLLDAPLLGRVGRTSRGLARLQLLETYSRR | |
RUO | |
NOXO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |