Learn More
Abnova™ Human NODAL Partial ORF (NP_060525, 275 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004838-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the TGF-beta superfamily. Studies of the mouse counterpart suggested that this gene may be essential for mesoderm formation and subsequent organization of axial structures in early embryonic development. [provided by RefSeq]
Sequence: RCEGECPNPVGEEFHPTNHAYIQSLLKRYQPHRVPSTCCAPVKTKPLSMLYVDNGRVLLDHHKDMIVEECGCSpecifications
NP_060525 | |
Liquid | |
4838 | |
NODAL (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC138230 | |
NODAL | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RCEGECPNPVGEEFHPTNHAYIQSLLKRYQPHRVPSTCCAPVKTKPLSMLYVDNGRVLLDHHKDMIVEECGC | |
RUO | |
NODAL | |
Wheat Germ (in vitro) | |
GST |