missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NNAT Partial ORF (NP_005377.1, 22 a.a. - 81 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005377.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4826 |
Molecular Weight (g/mol) | 32.34kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101955
|
Abnova™
H00004826-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16191945
|
Abnova™
H00004826-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a proteolipid that may be involved in the regulation of ion channels during brain development. The encoded protein may also play a role in forming and maintaining the structure of the nervous system. This gene is found within an intron of the BLCAP gene, but on the opposite strand. This gene is imprinted and is expressed only from the paternal allele, while BLCAP is not imprinted. Two transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]
Sequence: LLQVFLECCIYWVGFAFRNPPGTQPIARSEVFRYSLQKLAYTVSRTGRQVLGERRQRAPNSpecifications
NP_005377.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.34kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC1439/Peg5 | |
NNAT | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4826 | |
NNAT (Human) Recombinant Protein (Q01) | |
LLQVFLECCIYWVGFAFRNPPGTQPIARSEVFRYSLQKLAYTVSRTGRQVLGERRQRAPN | |
RUO | |
NNAT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |