missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NLRP2 Full-length ORF (AAH39269, 1 a.a. - 846 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH39269 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55655 |
Molecular Weight (g/mol) | 118.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101643
|
Abnova™
H00055655-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16191633
|
Abnova™
H00055655-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
NALP proteins, such as NALP2, are characterized by an N-terminal pyrin (MIM 608107) domain (PYD) and are involved in the activation of caspase-1 (CASP1; MIM 147678) by Toll-like receptors (see TLR4; MIM 603030). They may also be involved in protein complexes that activate proinflammatory caspases (Tschopp et al., 2003 [PubMed 12563287]).[supplied by OMIM]
Sequence: MVSSAQMGFNLQALLEQLSQDELSKFKYLITTFSLAHELQKIPHKEVDKADGKQLVEVLTTHCDSYWVEMASLQVFEKMHRMDLSERAKDEVREAALKSFNKRKPLSLGITRKERPPLDVDEMLERFKTEAQAFTETKGNVICLGKEVFKGKKPDKDNRCRYILKTKFREMWKSWPGDSKEVQVMAERYKMLIPFSNPRVLPGPFSYTVVLYGPAGLGKTTLAQKLMLDWAEDNLIHKFKYAFYLSCRELSRLGPCSFAELVFRDWPELQDDIPHILAQARKILFVIDGFDELGAAPGALIEDICGDWEKKKPVPVLLGSLLNRVMLPKAALLVTTRPRALRDLRILAEEPIYIRVEGFLEEDRRAYFLRHFGDEDQAMRAFELMRSNAALFQLGSAPAVCWIVCTTLKLQMEKGEDPVPTCLTRTGLFLRFLCSRFPQGAQLRGALRTLSLLAAQGLWAQTSVLHREDLERLGVQESDLRLFLDGDILRQDRVSKGCYSFIHLSFQQFLTALFYTLEKEEEEDRDGHTWDIGDVQKLLSGVERLRNPDLIQAGYYSFGLANEKRAKELEATFGCRMSPDIKQELLRCDISCKGGHSTVTDLQELLGCLYESQEEELVKEVMAQFKEISLHLNAVDVVPSSFCVKHCRNLQKMSLQVIKENLPENVTASESDAEVERSQDDQHMLPFWTDLCSIFGSNKDLMGLAINDSFLSASLVRILCEQIASDTCHLQRVVFKNISPADAHRNLCLALRGHKTVTYLTLQGNDQDDMFPALCEVLRHPECNLRYLGLVSCSATTQQWADLSLALEVNQSLTCVNLSDNELLDEGAKLLYTTLRHPKCFLQRLSSpecifications
AAH39269 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVSSAQMGFNLQALLEQLSQDELSKFKYLITTFSLAHELQKIPHKEVDKADGKQLVEVLTTHCDSYWVEMASLQVFEKMHRMDLSERAKDEVREAALKSFNKRKPLSLGITRKERPPLDVDEMLERFKTEAQAFTETKGNVICLGKEVFKGKKPDKDNRCRYILKTKFREMWKSWPGDSKEVQVMAERYKMLIPFSNPRVLPGPFSYTVVLYGPAGLGKTTLAQKLMLDWAEDNLIHKFKYAFYLSCRELSRLGPCSFAELVFRDWPELQDDIPHILAQARKILFVIDGFDELGAAPGALIEDICGDWEKKKPVPVLLGSLLNRVMLPKAALLVTTRPRALRDLRILAEEPIYIRVEGFLEEDRRAYFLRHFGDEDQAMRAFELMRSNAALFQLGSAPAVCWIVCTTLKLQMEKGEDPVPTCLTRTGLFLRFLCSRFPQGAQLRGALRTLSLLAAQGLWAQTSVLHREDLERLGVQESDLRLFLDGDILRQDRVSKGCYSFIHLSFQQFLTALFYTLEKEEEEDRDGHTWDIGDVQKLLSGVERLRNPDLIQAGYYSFGLANEKRAKELEATFGCRMSPDIKQELLRCDISCKGGHSTVTDLQELLGCLYESQEEELVKEVMAQFKEISLHLNAVDVVPSSFCVKHCRNLQKMSLQVIKENLPENVTASESDAEVERSQDDQHMLPFWTDLCSIFGSNKDLMGLAINDSFLSASLVRILCEQIASDTCHLQRVVFKNISPADAHRNLCLALRGHKTVTYLTLQGNDQDDMFPALCEVLRHPECNLRYLGLVSCSATTQQWADLSLALEVNQSLTCVNLSDNELLDEGAKLLYTTLRHPKCFLQRLS | |
RUO | |
NLRP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55655 | |
NLRP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLR19.9/FLJ20510/NALP2/NBS1/PAN1/PYPAF2 | |
NLRP2 | |
Recombinant | |
wheat germ expression system |