missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NINJ1 Full-length ORF (NP_004139.1, 1 a.a. - 152 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004139.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4814 |
Molecular Weight (g/mol) | 42.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16110768
|
Abnova™
H00004814-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16100768
|
Abnova™
H00004814-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
The ninjurin protein is upregulated after nerve injury both in dorsal root ganglion neurons and in Schwann cells (Araki and Milbrandt, 1996 [PubMed 8780658]). It demonstrates properties of a homophilic adhesion molecule and promotes neurite outgrowth from primary cultured dorsal root ganglion neurons.[supplied by OMIM]
Sequence: MDSGTEEYELNGGLPPGTPGSPDASPARWGWRHGPINVNHYASKKSAAESMLDIALLMANASQLKAVVEQGPSFAFYVPLVVLISISLVLQIGVGVLLIFLVKYDLNNPDKHAKLDFLNNLATGLVFIIVVVNIFITAFGVQKPLMDMAPQQSpecifications
NP_004139.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDSGTEEYELNGGLPPGTPGSPDASPARWGWRHGPINVNHYASKKSAAESMLDIALLMANASQLKAVVEQGPSFAFYVPLVVLISISLVLQIGVGVLLIFLVKYDLNNPDKHAKLDFLNNLATGLVFIIVVVNIFITAFGVQKPLMDMAPQQ | |
RUO | |
NINJ1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4814 | |
NINJ1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NIN1/NINJURIN | |
NINJ1 | |
Recombinant | |
wheat germ expression system |