missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NDUFA2 Full-length ORF (NP_002479.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_002479.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4695 |
Molecular Weight (g/mol) | 37.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16168191
|
Abnova™
H00004695-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16158191
|
Abnova™
H00004695-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The NDUFA2 gene encodes one of the accessory subunits of complex I, the first and largest complex of the mitochondrial respiratory chain (Hoefs et al., 2008 [PubMed 18513682]). For a discussion of complex I, see MIM 516000.[supplied by OMIM]
Sequence: MAAAAASRGVGAKLGLREIRIHLCQRSPGSQGVRDFIEKRYVELKKANPDLPILIRECSDVQPKLWARYAFGQETNVPLNNFSADQVTRALENVLSGKASpecifications
NP_002479.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAAAASRGVGAKLGLREIRIHLCQRSPGSQGVRDFIEKRYVELKKANPDLPILIRECSDVQPKLWARYAFGQETNVPLNNFSADQVTRALENVLSGKA | |
RUO | |
NDUFA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4695 | |
NDUFA2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B8/CD14 | |
NDUFA2 | |
Recombinant | |
wheat germ expression system |