Learn More
Abnova™ Human MYO3A Full-length ORF (AAH45538.1, 1 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH45538.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 53904 |
Molecular Weight (g/mol) | 54kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130433
|
Abnova™
H00053904-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16120433
|
Abnova™
H00053904-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene belongs to the myosin superfamily. Myosins are actin-dependent motor proteins and are categorized into conventional myosins (class II) and unconventional myosins (classes I and III through XV) based on their variable C-terminal cargo-binding domains. Class III myosins, such as this one, have a kinase domain N-terminal to the conserved N-terminal motor domains and are expressed in photoreceptors. The protein encoded by this gene plays an important role in hearing in humans. Three different recessive, loss of function mutations in the encoded protein have been shown to cause nonsyndromic progressive hearing loss. Expression of this gene is highly restricted, with the strongest expression in retina and cochlea. [provided by RefSeq]
Sequence: MFPLIGKTIIFDNFPDPSDTWEITETIGKGTYGKVFKVLNKKNGQKAAVKILDPIHDIDEEIEAGYNILKALSDHPNVVRFYGIYFKKDKVNGDKLWLVLELCSGGSVTDLVKGFLKRGERMSEPLIAYILHEALMGLQHLHNNKTIHRDVKGNNILLTTEGGVKLVDFGVSAQLTSTRHRRNTSVGTPFWMAPEVIACEQQLDTTYDARCDTWSLGITAIELGDGDPPLADLHPMRALFKIPRSDDSpecifications
AAH45538.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54kDa | |
Glutathione Sepharose 4 Fast Flow | |
MFPLIGKTIIFDNFPDPSDTWEITETIGKGTYGKVFKVLNKKNGQKAAVKILDPIHDIDEEIEAGYNILKALSDHPNVVRFYGIYFKKDKVNGDKLWLVLELCSGGSVTDLVKGFLKRGERMSEPLIAYILHEALMGLQHLHNNKTIHRDVKGNNILLTTEGGVKLVDFGVSAQLTSTRHRRNTSVGTPFWMAPEVIACEQQLDTTYDARCDTWSLGITAIELGDGDPPLADLHPMRALFKIPRSDD | |
RUO | |
MYO3A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
53904 | |
MYO3A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB30 | |
MYO3A | |
Recombinant | |
wheat germ expression system |