missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYO15A Partial ORF (NP_057323.2, 3432 a.a. - 3530 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051168-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16166676

  • 545.00€ / 25µg

Estimated Shipment: 27-09-2022
to see stock.
Add to basket

Description

Description

This gene encodes an unconventional myosin. This protein differs from other myosins in that it has a long N-terminal extension preceding the conserved motor domain. Studies in mice suggest that this protein is necessary for actin organization in the hair cells of the cochlea. Mutations in this gene have been associated with profound, congenital, neurosensory, nonsyndromal deafness. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Read-through transcripts containing an upstream gene and this gene have been identified, but they are not thought to encode a fusion protein. Several alternatively spliced transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq]

Sequence: PCILAINHNGLNFLSTETHELMVKFPLKEIQSTRTQRPTANSSYPYVEIALGDVAAQRTLQLQLEQGLELCRVVAVHVENLLSAHEKRLTLPPSEITLL
Specifications

Specifications

NP_057323.2
Liquid
51168
MYO15A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DFNB3/DKFZp686N18198/FLJ17274/FLJ31311/MYO15
MYO15A
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PCILAINHNGLNFLSTETHELMVKFPLKEIQSTRTQRPTANSSYPYVEIALGDVAAQRTLQLQLEQGLELCRVVAVHVENLLSAHEKRLTLPPSEITLL
RUO
MYO15A
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers