Learn More
ACROBiosystems Human Mucin-1 / MUC-1 Protein, Fc Tag
323.90€ - 2073.01€
Specifications
Accession Number | AAI20976 |
---|---|
Molecular Weight (g/mol) | 40.8 kDa |
Source | Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession # AAI20976). |
Immunogen | Ser 33 - Gly 167 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15817337
|
ACROBiosystems
MU1-H5252-1MG |
1 mg |
2073.01€
1mg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
15807337
|
ACROBiosystems
MU1-H5252-100UG |
100 μg |
323.90€
100µg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
Description
Mucin-1 (MUC1) is also known as Tumor-associated epithelial membrane antigen (EMA), Polymorphic epithelial mucin (PEM), Peanut-reactive urinary mucin (PUM), PEMT, Krebs von den Lungen-6 (KL-6), CD antigen CD227, Episialin, H23AG. MUC1 is a glycoprotein with extensive O-linked glycosylation of its extracellular domain. Mucins line the apical surface of epithelial cells in the lungs, stomach, intestines, eyes and several other organs. Mucins protect the body from infection by pathogen binding to oligosaccharides in the extracellular domain, preventing the pathogen from reaching the cell surface. Except protective function by binding to pathogens, MUC1 also functions in a cell signaling capacity. Overexpression of MUC1 is often associated with colon, breast, ovarian, lung and pancreatic cancers.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.8 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAI20976 | |
Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession # AAI20976). | |
-20°C | |
Mucin-1 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
40.8 kDa | |
Ser 33 - Gly 167 | |
Mucin 1,MUC1,CD227,EMA,H23AG,KL-6,MAM6,MUC-1/SEC,MUC-1/X,MUC1/ZD,PEM,PEMT,PUM,CA 15-3,Episialin | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |