missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Mucin-1 / MUC-1 Protein, Fc Tag

Brand:  ACROBiosystems MU1-H5252-1MG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15817337

  • 1452.66€ / 1mg


Estimated Shipment: 20-10-2021
Add to basket

Description

Description

Mucin-1 (MUC1) is also known as Tumor-associated epithelial membrane antigen (EMA), Polymorphic epithelial mucin (PEM), Peanut-reactive urinary mucin (PUM), PEMT, Krebs von den Lungen-6 (KL-6), CD antigen CD227, Episialin, H23AG. MUC1 is a glycoprotein with extensive O-linked glycosylation of its extracellular domain. Mucins line the apical surface of epithelial cells in the lungs, stomach, intestines, eyes and several other organs. Mucins protect the body from infection by pathogen binding to oligosaccharides in the extracellular domain, preventing the pathogen from reaching the cell surface. Except protective function by binding to pathogens, MUC1 also functions in a cell signaling capacity. Overexpression of MUC1 is often associated with colon, breast, ovarian, lung and pancreatic cancers.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.8 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

AAI20976
HEK293
Human
40.8 kDa
1 mg
Ser 33 - Gly 167
Mucin 1,MUC1,CD227,EMA,H23AG,KL-6,MAM6,MUC-1/SEC,MUC-1/X,MUC1/ZD,PEM,PEMT,PUM,CA 15-3,Episialin
Unconjugated
50 mM Tris, 100 mM Glycine, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession # AAI20976).
-20°C
Mucin-1
Fc Tag
Videos
SDS
Documents
Certificates