Learn More
Abnova™ Human MTUS1 Full-length ORF (AAH33842, 1 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH33842 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57509 |
Molecular Weight (g/mol) | 52.14kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16162743
|
Abnova™
H00057509-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16152743
|
Abnova™
H00057509-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein which contains a C-terminal domain able to interact with the angiotension II (AT2) receptor and a large coiled-coil region allowing dimerization. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. One of the transcript variants has been shown to encode a mitochondrial protein that acts as a tumor suppressor and partcipates in AT2 signaling pathways. Other variants may encode nuclear or transmembrane proteins but it has not been determined whether they also participate in AT2 signaling pathways. [provided by RefSeq]
Sequence: MQLQEQFDNLNAAHETSKLEIEASHSEKLELLTKAYEASLSEIKKGHEIEKKSLEDLLSEKQESLEKQINDLKSENDALNEKLKSEEQKRRAREKANLKNPQIMYLEQELESLKAVLEIKNEKLHQQDIKLMKMGKLVDNNTALVDKLKRFQQENEELKARMDKHMAISRQLSTEQAVLQESLEKESKVNKRLSMENEELLWKLHNGDLCSPKRSPTSSAIPLQSPRNSGSFPSPSISPRSpecifications
AAH33842 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.14kDa | |
Glutathione Sepharose 4 Fast Flow | |
MQLQEQFDNLNAAHETSKLEIEASHSEKLELLTKAYEASLSEIKKGHEIEKKSLEDLLSEKQESLEKQINDLKSENDALNEKLKSEEQKRRAREKANLKNPQIMYLEQELESLKAVLEIKNEKLHQQDIKLMKMGKLVDNNTALVDKLKRFQQENEELKARMDKHMAISRQLSTEQAVLQESLEKESKVNKRLSMENEELLWKLHNGDLCSPKRSPTSSAIPLQSPRNSGSFPSPSISPR | |
RUO | |
MTUS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
57509 | |
MTUS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATIP/DKFZp586D1519/DKFZp686F20243/FLJ14295/KIAA1288/MP44/MTSG1 | |
MTUS1 | |
Recombinant | |
wheat germ expression system |