missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MRPS5 Full-length ORF (NP_114108.1, 1 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00064969-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193833

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S5P family. Pseudogenes corresponding to this gene are found on chromosomes 4q, 5q, and 18q. [provided by RefSeq]

Sequence: MATAVRAVGCLPVLCSGTAGHLLGRQCSLNTLPAASILAWKSVLGNGHLSSLGTRDTHPYASLSRALQTQCCISSPSHLMSQQYRPYSFFTKLTADELWKGALAETGAGAKKGRGKRTKKKKRKDLNRGQIIGEGRYGFLWPGLNVPLMKNGAVQTIAQRSKEEQEKVEADMIQQREEWDRKKKMKVKRERGWSGNSWGGISLGPPDPGPCGETYEDFDTRILEVRNVFTMTAKEGRKKSIRVLVAVGNGKGAAGFSIGKATDRMDAFRKAKNRAVHHLHYIERYEDHTIFHDISLRFKRTHIKMKKQPKGYGLRCHRAIITICRLIGIKDMYAKVSGSINMLSLTQGLFRGLSRQETHQQLADKKGLHVVEIREECGPLPIVVASPRGPLRKDPEPEDEVPDVKLDWEDVKTAQGMKRSVWSNLKRAAT
Specifications

Specifications

NP_114108.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MRP-S5/S5mt
MRPS5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
64969
MRPS5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATAVRAVGCLPVLCSGTAGHLLGRQCSLNTLPAASILAWKSVLGNGHLSSLGTRDTHPYASLSRALQTQCCISSPSHLMSQQYRPYSFFTKLTADELWKGALAETGAGAKKGRGKRTKKKKRKDLNRGQIIGEGRYGFLWPGLNVPLMKNGAVQTIAQRSKEEQEKVEADMIQQREEWDRKKKMKVKRERGWSGNSWGGISLGPPDPGPCGETYEDFDTRILEVRNVFTMTAKEGRKKSIRVLVAVGNGKGAAGFSIGKATDRMDAFRKAKNRAVHHLHYIERYEDHTIFHDISLRFKRTHIKMKKQPKGYGLRCHRAIITICRLIGIKDMYAKVSGSINMLSLTQGLFRGLSRQETHQQLADKKGLHVVEIREECGPLPIVVASPRGPLRKDPEPEDEVPDVKLDWEDVKTAQGMKRSVWSNLKRAAT
RUO
MRPS5
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers