Learn More
Abnova™ Human MRPS22 Full-length ORF (AAH09296, 1 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH09296 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56945 |
Molecular Weight (g/mol) | 65.34kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102373
|
Abnova™
H00056945-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16192363
|
Abnova™
H00056945-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that does not seem to have a counterpart in prokaryotic and fungal-mitochondrial ribosomes. This gene lies telomeric of and is transcribed in the opposite direction from the forkhead box L2 gene. A pseudogene corresponding to this gene is found on chromosome Xq. [provided by RefSeq]
Sequence: MAPLGTTVLLWSLLRSSPGVERVCFRARIQPWHGGLLQPLPCSFEMGLPRRRFSSEAAESGSPETKKPTFMDEEVQSILTKMTGLNLQKTFKPAIQELKPPTYKLMTQAQLEEATRQAVEAAKVRLKMPPVLEERVPINDVLAEDKILEGTETTKYVFTDISYSIPHRERFIVVREPSGTLRKASWEERDRMIQVYFPKEGRKILTPIIFKEENLRTMYSQDRHVDVLNLCFAQFEPDSTEYIKVHHKTYEDIDKRGKYDLLRSTRYFGGMVWYFVNNKKIDGLLIDQIQRDLIDDATNLVQLYHVLHPDGQSAQGAKDQAAEGINLIKVFAKTEAQKGAYIELTLQTYQEALSRHSAASSpecifications
AAH09296 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.34kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAPLGTTVLLWSLLRSSPGVERVCFRARIQPWHGGLLQPLPCSFEMGLPRRRFSSEAAESGSPETKKPTFMDEEVQSILTKMTGLNLQKTFKPAIQELKPPTYKLMTQAQLEEATRQAVEAAKVRLKMPPVLEERVPINDVLAEDKILEGTETTKYVFTDISYSIPHRERFIVVREPSGTLRKASWEERDRMIQVYFPKEGRKILTPIIFKEENLRTMYSQDRHVDVLNLCFAQFEPDSTEYIKVHHKTYEDIDKRGKYDLLRSTRYFGGMVWYFVNNKKIDGLLIDQIQRDLIDDATNLVQLYHVLHPDGQSAQGAKDQAAEGINLIKVFAKTEAQKGAYIELTLQTYQEALSRHSAAS | |
RUO | |
MRPS22 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
56945 | |
MRPS22 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C3orf5/COXPD5/GIBT/GK002/MRP-S22/RPMS22 | |
MRPS22 | |
Recombinant | |
wheat germ expression system |