missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MPP5 Full-length ORF (NP_071919.2, 1 a.a. - 675 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00064398-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193583

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like (see DLG1; MIM 601014), ZO1-like (see TJP1; MIM 601009), p55-like (see MPP1; MIM 305360), and LIN2-like (see CASK; MIM 300172), based on their size and the presence of additional domains (Tseng et al., 2001 [PubMed 11311936]). MPP5 is a member of the p55-like MAGUK subfamily.[supplied by OMIM]

Sequence: MTTSHMNGHVTEESDSEVKNVDLASPEEHQKHREMAVDCPGDLGTRMMPIRRSAQLERIRQQQEDMRRRREEEGKKQELDLNSSMRLKKLAQIPPKTGIDNPMFDTEEGIVLESPHYAVKILEIEDLFSSLKHIQHTLVDSQSQEDISLLLQLVQNKDFQNAFKIHNAITVHMNKASPPFPLISNAQDLAQEVQTVLKPVHHKEGQELTALLNTPHIQALLLAHDKVAEQEMQLEPITDERVYESIGQYGGETVKIVRIEKARDIPLGATVRNEMDSVIISRIVKGGAAEKSGLLHEGDEVLEINGIEIRGKDVNEVFDLLSDMHGTLTFVLIPSQQIKPPPAKETVIHVKAHFDYDPSDDPYVPCRELGLSFQKGDILHVISQEDPNWWQAYREGDEDNQPLAGLVPGKSFQQQREAMKQTIEEDKEPEKSGKLWCAKKNKKKRKKVLYNANKNDDYDNEEILTYEEMSLYHQPANRKRPIILIGPQNCGQNELRQRLMNKEKDRFASAVPHTTRSRRDQEVAGRDYHFVSRQAFEADIAAGKFIEHGEFEKNLYGTSIDSVRQVINSGKICLLSLRTQSLKTLRNSDLKPYIIFIAPPSQERLRALLAKEGKNPKPEELREIIEKTREMEQNNGHYFDTAIVNSDLDKAYQELLRLINKLDTEPQWVPSTWLR
Specifications

Specifications

NP_071919.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
103.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12615/PALS1
MPP5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
64398
MPP5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTTSHMNGHVTEESDSEVKNVDLASPEEHQKHREMAVDCPGDLGTRMMPIRRSAQLERIRQQQEDMRRRREEEGKKQELDLNSSMRLKKLAQIPPKTGIDNPMFDTEEGIVLESPHYAVKILEIEDLFSSLKHIQHTLVDSQSQEDISLLLQLVQNKDFQNAFKIHNAITVHMNKASPPFPLISNAQDLAQEVQTVLKPVHHKEGQELTALLNTPHIQALLLAHDKVAEQEMQLEPITDERVYESIGQYGGETVKIVRIEKARDIPLGATVRNEMDSVIISRIVKGGAAEKSGLLHEGDEVLEINGIEIRGKDVNEVFDLLSDMHGTLTFVLIPSQQIKPPPAKETVIHVKAHFDYDPSDDPYVPCRELGLSFQKGDILHVISQEDPNWWQAYREGDEDNQPLAGLVPGKSFQQQREAMKQTIEEDKEPEKSGKLWCAKKNKKKRKKVLYNANKNDDYDNEEILTYEEMSLYHQPANRKRPIILIGPQNCGQNELRQRLMNKEKDRFASAVPHTTRSRRDQEVAGRDYHFVSRQAFEADIAAGKFIEHGEFEKNLYGTSIDSVRQVINSGKICLLSLRTQSLKTLRNSDLKPYIIFIAPPSQERLRALLAKEGKNPKPEELREIIEKTREMEQNNGHYFDTAIVNSDLDKAYQELLRLINKLDTEPQWVPSTWLR
RUO
MPP5
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers