Learn More
Abnova™ Human MNT Partial ORF (NP_064706, 473 a.a. - 582 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_064706 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4335 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16157891
|
Abnova™
H00004335-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16167891
|
Abnova™
H00004335-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
The Myc/Max/Mad network comprises a group of transcription factors that co-interact to regulate gene-specific transcriptional activation or repression. This gene encodes a protein member of the Myc/Max/Mad network. This protein has a basic-Helix-Loop-Helix-zipper domain (bHLHzip) with which it binds the canonical DNA sequence CANNTG, known as the E box, following heterodimerization with Max proteins. This protein is likely a transcriptional repressor and an antagonist of Myc-dependent transcriptional activation and cell growth. This protein represses transcription by binding to DNA binding proteins at its N-terminal Sin3-interaction domain. [provided by RefSeq]
Sequence: SAPSPAVQLAPATPPIGHITVHPATLNHVAHLGSQLPLYPQPVAVSHIAHTLSHQQVNGTAGLGPPATVMAKPAVGAQVVHHPQLVGQTVLNPVTMVTMPSFPVSTLKLASpecifications
NP_064706 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAD6/MXD6/ROX/bHLHd3 | |
MNT | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4335 | |
MNT (Human) Recombinant Protein (Q01) | |
SAPSPAVQLAPATPPIGHITVHPATLNHVAHLGSQLPLYPQPVAVSHIAHTLSHQQVNGTAGLGPPATVMAKPAVGAQVVHHPQLVGQTVLNPVTMVTMPSFPVSTLKLA | |
RUO | |
MNT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |