missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MMP21 Partial ORF (NP_671724.1, 374 a.a. - 481 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_671724.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 118856 |
Molecular Weight (g/mol) | 37.62kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16161607
|
Abnova™
H00118856-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16151607
|
Abnova™
H00118856-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the matrix metalloproteinase family. Proteins in this family are involved in the breakdown of extracellular matrix for both normal physiological processes, such as embryonic development, reproduction, and tissue remodeling,and disease processes, such as asthma and metastasis. The encoded protein may play an important role in embryogenesis, particularly in neuronal cells, as well as in lymphocyte development and survival. [provided by RefSeq]
Sequence: TRYGDPIQILTGWPGIPTHNIDAFVHIWTWKRDERYFFQGNQYWRYDSDKDQALTEDEQGKSYPKLISEGFPGIPSPLDTAFYDRRQKLIYFFKESLVFAFDVNRNRVSpecifications
NP_671724.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MMP21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
118856 | |
MMP21 (Human) Recombinant Protein (Q01) | |
TRYGDPIQILTGWPGIPTHNIDAFVHIWTWKRDERYFFQGNQYWRYDSDKDQALTEDEQGKSYPKLISEGFPGIPSPLDTAFYDRRQKLIYFFKESLVFAFDVNRNRV | |
RUO | |
MMP21 | |
Recombinant | |
wheat germ expression system |