missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MGC42638 Partial ORF (AAH40290, 1 a.a. - 75 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH40290 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 340561 |
Molecular Weight (g/mol) | 33.99kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112827
|
Abnova™
H00340561-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16102827
|
Abnova™
H00340561-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Sequence: MALKLIHKEFLELARDPQPHCSAGPVWDDMLHWQATITRPNDSSYLGGVFFLKFPSDYLFKPPKIKFTNGIYHQRSpecifications
AAH40290 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.99kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC42638 | |
UBE2DNL | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
340561 | |
MGC42638 (Human) Recombinant Protein (Q01) | |
MALKLIHKEFLELARDPQPHCSAGPVWDDMLHWQATITRPNDSSYLGGVFFLKFPSDYLFKPPKIKFTNGIYHQR | |
RUO | |
UBE2DNL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |