missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MCMDC1 Partial ORF (AAH31658, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00254394-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102607

  • 349.00€ / 10µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

This gene encodes a protein that shares similarity with minichromosome maintenance (MCM) proteins, which are known to be essential for initiation of DNA replication. [provided by RefSeq]

Sequence: MNSDQVTLVGQVFESYVSEYHKNDILLILKERDEDAHYPVVVNAMTLFETNMEIGEYFNMFPSEVLTIFDSALRRSALTILQSLSQPEAVSMKQNLHARI
Specifications

Specifications

AAH31658
Liquid
254394
MCMDC1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf61/FLJ13942/FLJ20170/FLJ56845/MCMDC1/MGC35304/dJ329L24.1/dJ329L24.3
MCM9
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MNSDQVTLVGQVFESYVSEYHKNDILLILKERDEDAHYPVVVNAMTLFETNMEIGEYFNMFPSEVLTIFDSALRRSALTILQSLSQPEAVSMKQNLHARI
RUO
MCM9
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers