missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MCCC1 Full-length ORF (AAH04214.1, 1 a.a. - 725 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH04214.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56922 |
Molecular Weight (g/mol) | 106.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16132343
|
Abnova™
H00056922-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16122343
|
Abnova™
H00056922-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
This gene encodes the large subunit of 3-methylcrotonyl-CoA carboxylase. This enzyme functions as a heterodimer and catalyzes the carboxylation of 3-methylcrotonyl-CoA to form 3-methylglutaconyl-CoA. Mutations in this gene are associated with 3-Methylcrotonylglycinuria, an autosomal recessive disorder of leucine catabolism. [provided by RefSeq]
Sequence: MAAASAVSVLLVAAERNRWHRLPSLLLPPRTWVWRQRTMKYTTATGRNITKVLIANRGEIACRVMRTAKKLGVQTVAVYSEADRNSMHVDMADEAYSIGPAPSQQSYLSMEKIIQVAKTSAAQAIHPGCGFLSENMEFAELCKQEGIIFIGPPPSAIRDMGIKSTSKSIMAAAGVPVVEGYHGEDQSDQCLKEHARRIGYPVMIKAVRGGGGKGMRIVRSEQEFQEQLESARREAKKSFNDDAMLIEKFVDTPRHVEVQVFGDHHGNAVYLFERDCSVQRRHQKIIEEAPAPGIKSEVRKKLGEAAVRAAKAVNYVGAGTVEFIMDSKHNFCFMEMNTRLQVEHPVTEMITGTDLVEWQLRIAAGEKIPLSQEEITLQGHAFEARIYAEDPSNNFMPVAGPLVHLSTPRADPSTRIETGVRQGDEVSVHYDPMIAKLVVWAADRQAALTKLRYSLRQYNIVGLPTNIDFLLNLSGHPEFEAGNVHTDFIPQHHKQLLLSRKAAAKESLCQAALGLILKEKAMTDTFTLQAHDQFSPFSSSSGRRLNISYTRNMTLKDGKNNVAIAVTYNHDGSYSMQIEDKTFQVLGNLYSEGDCTYLKCSVNGVASKAKLIILENTIYLFSKEGSIEIDIPVPKYLSSVSSQETQGGPLAPMTGTIEKVFVKAGDKVKAGDSLMVMIAMKMEHTIKSPKDGTVKKVFYREGAQANRHTPLVEFEEEESDKRESESpecifications
AAH04214.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
106.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAASAVSVLLVAAERNRWHRLPSLLLPPRTWVWRQRTMKYTTATGRNITKVLIANRGEIACRVMRTAKKLGVQTVAVYSEADRNSMHVDMADEAYSIGPAPSQQSYLSMEKIIQVAKTSAAQAIHPGCGFLSENMEFAELCKQEGIIFIGPPPSAIRDMGIKSTSKSIMAAAGVPVVEGYHGEDQSDQCLKEHARRIGYPVMIKAVRGGGGKGMRIVRSEQEFQEQLESARREAKKSFNDDAMLIEKFVDTPRHVEVQVFGDHHGNAVYLFERDCSVQRRHQKIIEEAPAPGIKSEVRKKLGEAAVRAAKAVNYVGAGTVEFIMDSKHNFCFMEMNTRLQVEHPVTEMITGTDLVEWQLRIAAGEKIPLSQEEITLQGHAFEARIYAEDPSNNFMPVAGPLVHLSTPRADPSTRIETGVRQGDEVSVHYDPMIAKLVVWAADRQAALTKLRYSLRQYNIVGLPTNIDFLLNLSGHPEFEAGNVHTDFIPQHHKQLLLSRKAAAKESLCQAALGLILKEKAMTDTFTLQAHDQFSPFSSSSGRRLNISYTRNMTLKDGKNNVAIAVTYNHDGSYSMQIEDKTFQVLGNLYSEGDCTYLKCSVNGVASKAKLIILENTIYLFSKEGSIEIDIPVPKYLSSVSSQETQGGPLAPMTGTIEKVFVKAGDKVKAGDSLMVMIAMKMEHTIKSPKDGTVKKVFYREGAQANRHTPLVEFEEEESDKRESE | |
RUO | |
MCCC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
56922 | |
MCCC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686B20267/FLJ25545/MCC-B/MCCA | |
MCCC1 | |
Recombinant | |
wheat germ expression system |