missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAP3K13 Partial ORF (NP_004712.1, 711 a.a. - 810 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004712.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9175 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16110096
|
Abnova™
H00009175-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16100096
|
Abnova™
H00009175-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a member of serine/threonine protein kinase family. This kinase contains a dual leucine-zipper motif, and has been shown to form dimers/oligomers through its leucine-zipper motif. This kinase can phosphorylate and activate MAPK8/JNK, MAP2K7/MKK7, which suggests a role in the JNK signaling pathway. [provided by RefSeq]
Sequence: SEPDKGQAGPWGCCQADAYDPCLQCRPEQYGSLDIPSAEPVGRSPDLSKSPAHNPLLENAQSSEKTEENEFSGCRSESSLGTSHLGTPPALPRKTRPLQKSpecifications
NP_004712.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LZK/MGC133196 | |
MAP3K13 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9175 | |
MAP3K13 (Human) Recombinant Protein (Q01) | |
SEPDKGQAGPWGCCQADAYDPCLQCRPEQYGSLDIPSAEPVGRSPDLSKSPAHNPLLENAQSSEKTEENEFSGCRSESSLGTSHLGTPPALPRKTRPLQK | |
RUO | |
MAP3K13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |