missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAGEC2 Partial ORF (NP_057333.1, 146 a.a. - 222 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_057333.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51438 |
Molecular Weight (g/mol) | 34.21kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16156976
|
Abnova™
H00051438-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16146976
|
Abnova™
H00051438-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
This gene is related to members of the MAGEC gene family. It is not expressed in normal tissues, except for testis, and is expressed in tumors of various histological types. This gene and the MAGEC genes are clustered on chromosome Xq26-q27. [provided by RefSeq]
Sequence: AELVEFLLLKYEAEEPVTEAEMLMIVIKYKDYFPVILKRAREFMELLFGLALIEVGPDHFCVFANTVGLTDEGSDDESpecifications
NP_057333.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.21kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CT10/HCA587/MAGEE1/MGC13377 | |
MAGEC2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
51438 | |
MAGEC2 (Human) Recombinant Protein (Q01) | |
AELVEFLLLKYEAEEPVTEAEMLMIVIKYKDYFPVILKRAREFMELLFGLALIEVGPDHFCVFANTVGLTDEGSDDE | |
RUO | |
MAGEC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |