missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LRRFIP2 Full-length ORF (NP_060194.1, 1 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_060194.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9209 |
Molecular Weight (g/mol) | 71.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16133402
|
Abnova™
H00009209-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16123402
|
Abnova™
H00009209-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
Sequence: MGTPASGRKRTPVKDRFSAEDEALSNIAREAEARLAAKRAARAEARDIRMRELERQQKELDEKSDKQYAENYTRPSSRNSASATTPLSGNSSRRGSGDTSSLIDPDTSLSELRESLSEVEEKYKKAMVSNAQLDNEKNNLIYQVDTLKDVIEEQEEQMAEFYRENEEKSKELERQKHMCSVLQHKMEELKEGLRQRDELIEKHGLVIIPDGTPNGDVSHEPVAGAITVVSQEAAQVLESAGEGPLDVRLRKLAGEKEELLSQIRKLKLQLEEERQKCSRNDGTVGDLAGLQNGSDLQFIEMQRDANRQISEYKFKLSKAEQDITTLEQSISRLEGQVLRYKTAAENAEKVEDELKAEKRKLQRELRTALDKIEEMEMTNSHLAKRLEKMKANRTALLAQQSpecifications
NP_060194.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGTPASGRKRTPVKDRFSAEDEALSNIAREAEARLAAKRAARAEARDIRMRELERQQKELDEKSDKQYAENYTRPSSRNSASATTPLSGNSSRRGSGDTSSLIDPDTSLSELRESLSEVEEKYKKAMVSNAQLDNEKNNLIYQVDTLKDVIEEQEEQMAEFYRENEEKSKELERQKHMCSVLQHKMEELKEGLRQRDELIEKHGLVIIPDGTPNGDVSHEPVAGAITVVSQEAAQVLESAGEGPLDVRLRKLAGEKEELLSQIRKLKLQLEEERQKCSRNDGTVGDLAGLQNGSDLQFIEMQRDANRQISEYKFKLSKAEQDITTLEQSISRLEGQVLRYKTAAENAEKVEDELKAEKRKLQRELRTALDKIEEMEMTNSHLAKRLEKMKANRTALLAQQ | |
RUO | |
LRRFIP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9209 | |
LRRFIP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434H2035/FLJ20248/FLJ22683/FLJ58304/HUFI-2 | |
LRRFIP2 | |
Recombinant | |
wheat germ expression system |