missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LRP1B Partial ORF (NP_061027, 111 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_061027 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 53353 |
Molecular Weight (g/mol) | 35.42kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16117396
|
Abnova™
H00053353-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16127396
|
Abnova™
H00053353-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
LRP1B belongs to the low density lipoprotein (LDL) receptor gene family. These receptors play a wide variety of roles in normal cell function and development due to their interactions with multiple ligands (Liu et al., 2001 [PubMed 11384978]).[supplied by OMIM]
Sequence: VHCQELLSNCQQLNCQYKCTMVRNSTRCYCEDGFEITEDGRSCKDQDECAVYGTCSQTCRNTHGSYTCSCVEGYLMQPDNRSCKAKIEPTSpecifications
NP_061027 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LRP-DIT/LRPDIT | |
LRP1B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
53353 | |
LRP1B (Human) Recombinant Protein (Q01) | |
VHCQELLSNCQQLNCQYKCTMVRNSTRCYCEDGFEITEDGRSCKDQDECAVYGTCSQTCRNTHGSYTCSCVEGYLMQPDNRSCKAKIEPT | |
RUO | |
LRP1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |