missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LOC90321 Full-length ORF (NP_001010851.1, 1 a.a. - 468 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001010851.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 90321 |
Molecular Weight (g/mol) | 80.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16116473
|
Abnova™
H00090321-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16106473
|
Abnova™
H00090321-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
Sequence: MAQLRRGHLTFRDVAIEFSQEEWKCLDPVQKALYRDVMLENYRNLVSLGICLPDLSIISMMKQRTEPWTVENEMKVAKNPDRWEGIKDINTGRSCAVRSKAGNKPITNQLGLTFQLPLPELEIFQGEGKIYECNQVQKFISHSSSVSPLQRIYSGVKTHIFNKHRNDFVDFPLLSQEQKAHIRRKPYECNEQGKVFRVSSSLPNHQVIHTADKPNRCHECGKTVRDKSGLAEHWRIRTGEKPYKCKECGKLFNRIAYLARHEKVHTGESPYKCNECGKVFSRITYLVRHQKIHTREKPHKCNKCGKVYSSSSYLAQHWRIHTGEKLYKCNKCGKEFSGHSSLTTHLLIHTGEKPYKCKECDKAFRHKFSLTVHQRNHNGEKPYKCHECGKVFTQVSHLARHQKIHTGEKPYKCNECGKVFTQNSHLANHQRIHTGEKPYKCHVCGKVFRHSSWFVQHQRSVHERVLTNSpecifications
NP_001010851.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAQLRRGHLTFRDVAIEFSQEEWKCLDPVQKALYRDVMLENYRNLVSLGICLPDLSIISMMKQRTEPWTVENEMKVAKNPDRWEGIKDINTGRSCAVRSKAGNKPITNQLGLTFQLPLPELEIFQGEGKIYECNQVQKFISHSSSVSPLQRIYSGVKTHIFNKHRNDFVDFPLLSQEQKAHIRRKPYECNEQGKVFRVSSSLPNHQVIHTADKPNRCHECGKTVRDKSGLAEHWRIRTGEKPYKCKECGKLFNRIAYLARHEKVHTGESPYKCNECGKVFSRITYLVRHQKIHTREKPHKCNKCGKVYSSSSYLAQHWRIHTGEKLYKCNKCGKEFSGHSSLTTHLLIHTGEKPYKCKECDKAFRHKFSLTVHQRNHNGEKPYKCHECGKVFTQVSHLARHQKIHTGEKPYKCNECGKVFTQNSHLANHQRIHTGEKPYKCHVCGKVFRHSSWFVQHQRSVHERVLTN | |
RUO | |
ZNF766 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
90321 | |
LOC90321 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ZNF766 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |