missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LILRA2 Partial ORF (NP_006857, 322 a.a. - 383 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011027-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16103546

  • 349.00€ / 10µg

Estimated Shipment: 06-07-2023
to see stock.
Add to basket

Description

Description

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (see, e.g., LILRB1, MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades. One member of subfamily A (LILRA3; MIM 604818) lacks a transmembrane region and is presumed to be a soluble receptor.[supplied by OMIM]

Sequence: DRPSLSVQPVPTVAPGKNVTLLCQSRGQFHTFLLTKEGAGHPPLHLRSEHQAQQNQAEFRMG
Specifications

Specifications

NP_006857
Liquid
11027
LILRA2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD85H/ILT1/LIR-7/LIR7
LILRA2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.56kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DRPSLSVQPVPTVAPGKNVTLLCQSRGQFHTFLLTKEGAGHPPLHLRSEHQAQQNQAEFRMG
RUO
LILRA2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers