missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human LAG3 / CD223 Protein, Fc Tag

MALS verified

Brand:  ACROBiosystems LA3-H5255-100UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15817217

  • 302.64€ / 100µg


Estimated Shipment: 25-10-2021
Add to basket

Description

Description

Lymphocyte activation gene 3 protein (LAG3) is also known as CD antigen CD223 and protein FDC, which belongs to immunoglobulin (Ig) superfamily and contains 4 extracellular Ig-like domains. The LAG3 gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG3 to CD4. LAG3 /CD223 involved in lymphocyte activation. LAG3 /CD223 binds to HLA class-II antigens.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.8 kDa. The protein migrates as 80-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

NP_002277.4
HEK293
Human
72.8 kDa
100 µg
Leu 23 - Leu 450
LAG3,CD223,FDC
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>95% as determined by SDS-PAGE
Human LAG-3, Fc Tag (LA3-H5255) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Leu 450 (Accession # P18627-1).
-20°C
LAG-3
Fc Tag
Videos
SDS
Documents
Certificates