Learn More
Abnova™ Human KCNJ2 Partial ORF (NP_000882, 328 a.a. - 427 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_000882 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3759 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127101
|
Abnova™
H00003759-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16137101
|
Abnova™
H00003759-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, probably participates in establishing action potential waveform and excitability of neuronal and muscle tissues. Mutations in this gene have been associated with Andersen syndrome, which is characterized by periodic paralysis, cardiac arrhythmias, and dysmorphic features. [provided by RefSeq]
Sequence: PVLFEEKHYYKVDYSRFHKTYEVPNTPLCSARDLAEKKYILSNANSFCYENEVALTSKEEDDSENGVPESTSTDTPPDIDLHNQASVPLEPRPLRRESEISpecifications
NP_000882 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HHBIRK1/HHIRK1/IRK1/KIR2.1/LQT7/SQT3 | |
KCNJ2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3759 | |
KCNJ2 (Human) Recombinant Protein (Q01) | |
PVLFEEKHYYKVDYSRFHKTYEVPNTPLCSARDLAEKKYILSNANSFCYENEVALTSKEEDDSENGVPESTSTDTPPDIDLHNQASVPLEPRPLRRESEI | |
RUO | |
KCNJ2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |